
5-(2,4-Difluorophenyl)isoxazole
Catalog # APOSPC9964-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:138716-46-8
Formula:C₉H₅F₂NO
MDL Number:MFCD02183538