
(4-Chloro-3-fluorophenyl)acetic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:883500-51-4
:
Formula:(C₆H₃ClF)CH₂COOH
MDL Number:MFCD04115861
:188.59 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together