Order Entry
France
ContactUsLinkComponent
4-Amino-2,3-difluorophenol
Corrosive
Irritant
4-Amino-2,3-difluorophenol
Catalog # APOSPC8312-100MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-Amino-2,3-difluorophenol
Catalog # APOSPC8312-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :145.11 g/mol
Melting Pt:150…155 °C (dec.)
Storage Temperature:Freezer
UN:2928
ADR:6.1+8,II
 :GHS05
 :GHS07
CAS Number:163733-99-1
Formula:C₆H₅F₂NO
MDL Number:MFCD03094494