
(4-Chloro-2,6-difluorophenyl)acetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:537033-55-9
:
Formula:C₈H₅ClF₂O₂
MDL Number:MFCD03094467
Melting Pt:137…138 °C
:206.58 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together