Ethyl-6,8-difluoro-4-hydroxyquinoline-3-carboxylate
Catalog # APOSPC8078-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:107555-38-4
Formula:C₁₂H₉F₂NO
MDL Number:MFCD00173349