
2-Bromo-N,N-dimethyl-5-fluorobenzamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:951884-08-5
:
Formula:C₉H₉BrFNO
MDL Number:MFCD09800904
:246.0763 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together