
1H,1H,8H,8H-Octafluoro-3,6-dioxaoctane-1,8-diol
Catalog # APOSPC7617-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:103 °C/1.3mmHg
Melting Pt:67…68 °C
:294.1 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:129301-42-4
Formula:C₆H₆F₈O₄
MDL Number:MFCD03094435