
4,4,4-Trifluorocrotonamide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:590-76-1
:
Formula:C₄H₄F₃NO
MDL Number:MFCD00190644
:139.08 g/mol
Melting Pt:147…150 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together