2,4,6-Trifluorobenzyl bromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:151411-98-2
:
Formula:C₇H₄BrF₃
MDL Number:MFCD00061210
:225.01 g/mol
UN:3265
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together