

3,4,5-Trifluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:163733-96-8
:
Formula:C₆H₄F₃N
MDL Number:MFCD00083522
Melting Pt:61…64 °C
:147.1 g/mol
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together