
6,8-Difluoro-4-quinolinol
Catalog # APOSPC7213-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:243448-16-0
Formula:C₉H₅F₂NO
MDL Number:MFCD00278786