
2-Bromo-5-fluorocinnamic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:202865-70-1
:
Formula:C₉H₆BrFO₂
MDL Number:MFCD00142878
:245.05 g/mol
Melting Pt:188…192 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together