3-Fluoro-2-methylpyridine 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15931-15-4
:
Formula:C₆H₆FN
MDL Number:MFCD08705710
:111.119 g/mol
Boiling Pt:114 °C
Density:1.077 g/cm³ (25 °C)
Flash Pt:23 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together