

Hexafluoroglutaric anhydride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:376-68-1
:
Formula:C₅F₆O₃
MDL Number:MFCD00006680
:222.04 g/mol
Boiling Pt:72 °C
Density:1.654 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:206-811-8
UN:3265
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together