Order Entry
France
ContactUsLinkComponent
Diisopropyl-3-fluorocyclobutane-1,1-dicarboxylate
Diisopropyl-3-fluorocyclobutane-1,1-dicarboxylate
Catalog # APOSPC420006-1G
Supplier:  Apollo Scientific
CAS Number:  
Diisopropyl-3-fluorocyclobutane-1,1-dicarboxylate
Catalog # APOSPC420006-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.28 g/mol
Storage Temperature:Ambient
CAS Number:1403767-29-2
Formula:C₁₂H₁₉FO₄
MDL Number:MFCD23105821