
4-Fluorophthalic anhydride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:319-03-9
:
Formula:C₈H₃FO₃
MDL Number:MFCD00191363
:166.11 g/mol
Boiling Pt:147…149 °C/20mmHg
Melting Pt:74…79 °C
Density:1.5 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:923-092-2
:GHS07
Frequently Bought Together