Order Entry
France
ContactUsLinkComponent
(2-{[1-Ethyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methoxy}ethyl)amine hydrochloride
(2-{[1-Ethyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methoxy}ethyl)amine hydrochloride
Catalog # APOSPC410105-1G
Supplier:  Apollo Scientific
CAS Number:  
(2-{[1-Ethyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methoxy}ethyl)amine hydrochloride
Catalog # APOSPC410105-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :273.69 g/mol
Storage Temperature:Ambient
CAS Number:1417570-03-6
Formula:C₉H₁₅ClF₃N₃O
MDL Number:MFCD22421975