4-(3,4-Difluorophenyl)piperidine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:721958-67-4
:
Formula:C₁₁H₁₄ClF₂N
MDL Number:MFCD04037205
:233.69 g/mol
Storage Temperature:Ambient
Frequently Bought Together