5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride
Catalog # APOSPC401009-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.6 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:175277-48-2
Formula:C₇H₇ClF₃N₃
MDL Number:MFCD00203995