
4-Nitro-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid
Catalog # APOSPC400600-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.08 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1322805-15-1
Formula:C₅H₂F₃N₃O₄
MDL Number:MFCD11845715