Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-2-hydroxy-3,3,3-trifluoropropanoate
Methyl-2-hydroxy-3,3,3-trifluoropropanoate 95%
Methyl-2-hydroxy-3,3,3-trifluoropropanoate 95%
APOSPC3844-1G
:
Apollo Scientific
:
Methyl-2-hydroxy-3,3,3-trifluoropropanoate 95%
APOSPC3844-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
158.08 g/mol
:
60…64 °C/20mmHg
:
59…60 °C
:
GHS07
:
Ambient
:
93496-85-6
:
C₄H₅F₃O₃
:
MFCD02093316