Order Entry
France
ContactUsLinkComponent
3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carbonitrile
3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carbonitrile
Catalog # APOSPC32294-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carbonitrile
Catalog # APOSPC32294-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.24 g/mol
Storage Temperature:Ambient
CAS Number:317840-08-7
Formula:C₁₀H₆F₃N₃S
MDL Number:MFCD00486825