4-Fluorobenzoic anhydride 95+%
Catalog # APOSPC31419-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:262.21 g/mol
Storage Temperature:Ambient
CAS Number:25569-77-1
Formula:C₁₄H₈F₂O₃
MDL Number:MFCD00425938