
3-Chloro-5-(trifluoromethoxy)phenylacetic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:433938-70-6
:
Formula:C₉H₆ClF₃O₃
MDL Number:MFCD06660313
:254.59 g/mol
Melting Pt:42…44 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together