
2-Nitro-5-(trifluoromethoxy)benzoic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:189359-65-7
:
Formula:C₈H₄F₃NO₅
MDL Number:MFCD08741392
:251.12 g/mol
Melting Pt:97.8…98.5 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together