Order Entry
France
ContactUsLinkComponent
2,3-Difluoro-4-iodophenol 97%
Irritant
2,3-Difluoro-4-iodophenol 97%
Catalog # APOSPC2912-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Difluoro-4-iodophenol 97%
Catalog # APOSPC2912-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :255.99 g/mol
Melting Pt:53…63 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:144292-40-0
Formula:C₆H₃F₂IO
MDL Number:MFCD08741391