
4-Cyano-3-fluorobenzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:101048-76-4
:
Formula:C₈H₄FNO
MDL Number:MFCD06738989
:149.1200 g/mol
Melting Pt:86…90 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together