
2-Bromo-4,6-difluorophenol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98130-56-4
:
Formula:C₆H₃BrF₂O
MDL Number:MFCD00070752
:208.99 g/mol
Boiling Pt:153…154 °C
Density:1.821 g/cm³ (25 °C)
Flash Pt:105.6 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together