cis-3,4-Difluoropyrrolidine hydrochloride 95+%
Catalog # APOSPC201334-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (3S,4R)-rel-3,4-Difluoropyrrolidine hydrochloride
:
:143.56 g/mol
Storage Temperature:Ambient
CAS Number:869481-94-7
Formula:C₄H₈ClF₂N
MDL Number:MFCD20230545