6-[4-(Trifluoromethyl)piperidino]nicotinic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1033463-36-3
:
Formula:C₁₂H₁₃F₃N₂O₂
MDL Number:MFCD10758091
:274.24 g/mol
Storage Temperature:Ambient
Frequently Bought Together