
Methyl-2-chloro-4-(trifluoromethyl)nicotinate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:196708-48-2
:
Formula:C₈H₅ClF₃NO₂
MDL Number:MFCD12827815
:239.58 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together