
(2-Chloro-6-fluorophenyl)acetic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:37777-76-7
:
Formula:(C₆H₃ClF)CH₂COOH
MDL Number:MFCD00004319
:188.59 g/mol
Melting Pt:120…123 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together