(3-Chloro-2-fluorophenyl)acetic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261762-96-3
:
Formula:(C₆H₃ClF)CH₂COOH
MDL Number:MFCD01631554
:188.59 g/mol
Melting Pt:128…129 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together