Order Entry
France
ContactUsLinkComponent
2-(Boc-amino)-4-methylpyridine 95%
2-(Boc-amino)-4-methylpyridine 95%
Catalog # APOSOR965752-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(Boc-amino)-4-methylpyridine 95%
Catalog # APOSOR965752-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :208.261 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:90101-20-5
Formula:C₁₁H₁₆N₂O₂
MDL Number:MFCD07776933