Order Entry
France
ContactUsLinkComponent
Ethyl-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate 95%
Ethyl-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate 95%
Catalog # APOSOR963490-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate 95%
Catalog # APOSOR963490-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :185.2200 g/mol
Storage Temperature:Ambient
CAS Number:106556-66-5
Formula:C₉H₁₅NO₃
MDL Number:MFCD09835297