4-Bromo-3-methylbenzaldehyde 98%
Catalog # APOSOR959850-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:199.05 g/mol
Storage Temperature:Ambient
CAS Number:78775-11-8
Formula:C₈H₇BrO
MDL Number:MFCD04971197