Ethyl nicotinate 98%
Catalog # APOSOR954544-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:151.16 g/mol
Storage Temperature:Ambient
CAS Number:614-18-6
Formula:C₈H₉NO₂
MDL Number:MFCD00006389