
1H-Pyrrolo[3,2-b]pyridine-6-carbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1020056-33-0
:
Formula:C₈H₆N₂O
MDL Number:MFCD09859114
:146.15 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together