Order Entry
France
ContactUsLinkComponent
(4-Chloropyrazol-1-yl)acetic acid 95%
Irritant
(4-Chloropyrazol-1-yl)acetic acid 95%
Catalog # APOSOR943527-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(4-Chloropyrazol-1-yl)acetic acid 95%
Catalog # APOSOR943527-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :160.56 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:32089-46-6
Formula:C₅H₅ClN₂O₂
MDL Number:MFCD00036053