Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate 96%
Catalog # APOSOR936808-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: cis-Dimethyl cyclohex-4-ene-1,2-dicarboxylate, cis-4-Cyclohexene-1,2-dicarboxylic Acid Dimethyl Ester
:
:198.22 g/mol
Storage Temperature:Ambient
CAS Number:4841-84-3
Formula:C₁₀H₁₄O₄
MDL Number:MFCD00209581