Order Entry
France
ContactUsLinkComponent
Phenylpropiolic acid 95%
Phenylpropiolic acid 95%
Catalog # APOSOR931330-100G
Supplier:  Apollo Scientific
CAS Number:  
Phenylpropiolic acid 95%
Catalog # APOSOR931330-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :146.15 g/mol
Storage Temperature:Ambient
CAS Number:637-44-5
Formula:C₉H₆O₂
MDL Number:MFCD00004361