3-(4-Hydroxy-3-methoxyphenyl)propionic acid 97%
Catalog # APOSOR925053-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: Dihydroferulic acid, 3-(4-Hydroxy-3-methoxyphenyl)propanoic acid
:
:196.20 g/mol
Storage Temperature:Ambient
CAS Number:1135-23-5
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00016558