Order Entry
France
ContactUsLinkComponent
Propyl-4-hydroxybenzoate 95%
Propyl-4-hydroxybenzoate 95%
Catalog # APOSOR923454-100G
Supplier:  Apollo Scientific
CAS Number:  
Propyl-4-hydroxybenzoate 95%
Catalog # APOSOR923454-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :180.2 g/mol
Storage Temperature:Ambient
CAS Number:94-13-3
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00002354