Order Entry
France
ContactUsLinkComponent
1-(2-Methylpyridin-4-yl)ethanone 95%
1-(2-Methylpyridin-4-yl)ethanone 95%
Catalog # APOSOR902630-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Methylpyridin-4-yl)ethanone 95%
Catalog # APOSOR902630-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :135.1632 g/mol
Storage Temperature:Ambient
CAS Number:2732-28-7
Formula:C₈H₉NO
MDL Number:MFCD07375079