
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98-79-3
:
Formula:C₅H₇NO₃
MDL Number:MFCD00005272
:129.1 g/mol
Melting Pt:155…163 °C
EINECS:202-700-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together