3-Amino-2-methylpyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3430-10-2
:
Formula:C₆H₈N₂
MDL Number:MFCD03788195
:108.14 g/mol
Melting Pt:114…119 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together