2-Methyl-2-[(5-nitro-2-pyridinyl)sulphanyl]propanoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:952183-12-9
:
Formula:C₉H₁₀N₂O₄S
MDL Number:MFCD09607896
:242.26 g/mol
Melting Pt:60…62 °C
Storage Temperature:Ambient
Frequently Bought Together