3-Amino-5-methylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3430-19-1
:
Formula:C6H8N2
MDL Number:MFCD04112508
:108.1411 g/mol
Boiling Pt:155 °C/21mmHg
Melting Pt:59…65 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together