4-Chloroindole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25235-85-2
:
Formula:C₈H₆ClN
MDL Number:MFCD00005665
:151.6 g/mol
Boiling Pt:129…130 °C/4mmHg
Density:1.259 g/cm³ (25 °C)
Flash Pt:113 °C
EINECS:246-747-8
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together