2-Amino-3-bromo-6-methylpyridine 98%
Catalog # APOSOR6620-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:187.04 g/mol
Melting Pt:77…79 °C
:GHS07
Storage Temperature:Ambient
CAS Number:126325-46-0
Formula:C₆H₇BrN₂
MDL Number:MFCD03095215